Assay Details
| Gene Symbol: Pick1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045558.1 | NP_001039023.1 | 11 - 12 | 1091 | 66 | |
| NM_008837.3 | NP_032863.2 | 11 - 12 | 1150 | 66 | ||
| GenBank mRNA | AK090155.1 | - | 1083 | 66 | ||
| BC048788.1 | - | 11 - 12 | 1036 | 6488048 | 66 | |
| BE652516.1 | - | 3 - 4 | 286 | 66 | ||
| CD363874.1 | - | 8 - 9 | 580 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pick1
Genomic Map