Assay Details
| Gene Symbol: Pick1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008837.3 | NP_032863.2 | 1 - 2 | 248 | 94 | |
| GenBank mRNA | BU698586.1 | - | 1 - 2 | 247 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_008837.3 transcripts.
Set Membership: |
|

Pick1
Genomic Map