Assay Details
| Gene Symbol: Pick1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045558.1 | NP_001039023.1 | 3 - 4 | 402 | 75 | |
| NM_008837.3 | NP_032863.2 | 3 - 4 | 461 | 75 | ||
| GenBank mRNA | AK050224.1 | - | 3 - 4 | 373 | 75 | |
| AK090155.1 | - | 394 | 75 | |||
| BC023088.1 | - | 3 - 4 | 368 | 75 | ||
| BC048788.1 | - | 3 - 4 | 347 | 6488048 | 75 | |
| BU057774.1 | - | 3 - 4 | 364 | 75 | ||
| BU698586.1 | - | 3 - 4 | 460 | 75 | ||
| CA468666.1 | - | 3 - 4 | 282 | 75 | ||
| CK791908.1 | - | 3 - 4 | 402 | 75 | ||
| Z46720.1 | - | 2 - 3 | 150 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pick1
Genomic Map