Assay Details
| Gene Symbol: Tomm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172609.3 | NP_766197.2 | 1 - 2 | 138 | 74 | |
| GenBank mRNA | AK009868.1 | - | 1 - 2 | 138 | 74 | |
| AK013471.1 | - | 1 - 2 | 127 | 74 | ||
| AK049442.1 | - | 126 | 74 | |||
| AK150360.1 | - | 1 - 2 | 126 | 74 | ||
| AK150787.1 | - | 1 - 2 | 135 | 74 | ||
| AK152843.1 | - | 1 - 2 | 126 | 74 | ||
| AK167803.1 | - | 1 - 2 | 126 | 74 | ||
| BC056920.1 | - | 1 - 2 | 120 | 6819748 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tomm22
Genomic Map