Assay Details
| Gene Symbol: Maff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156896.1 | - | 1 - 2 | 212 | 88 | |
| BC022952.1 | - | 1 - 2 | 75 | 5132212 | 88 | |
| CX211309.1 | - | 1 - 2 | 233 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156896.1, BC022952.1 transcripts.
Set Membership: |
|

Maff
Genomic Map