Assay Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028634.3 | NP_082910.1 | 1 - 2 | 67 | 68 | |
| GenBank mRNA | AF331040.1 | - | 1 - 2 | 67 | 68 | |
| AK003719.1 | - | 1 - 2 | 43 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF331040.1, AK003719.1, NM_028634.3 transcripts.
Set Membership: |
|

Cby1
Genomic Map