Assay Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028634.3 | NP_082910.1 | 4 - 5 | 442 | 57 | |
| GenBank mRNA | AF331040.1 | - | 4 - 5 | 442 | 57 | |
| AK003719.1 | - | 4 - 5 | 418 | 57 | ||
| BC005733.1 | - | 3 - 4 | 395 | 3596486 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein localization
cell differentiation
negative regulation of Wnt signaling pathway
cilium assembly
fat cell differentiation
negative regulation of transcription, DNA-templated
cardiac muscle cell differentiation
negative regulation of canonical Wnt signaling pathway
beta-catenin binding
identical protein binding
protein localization
cell differentiation
negative regulation of Wnt signaling pathway
cilium assembly
fat cell differentiation
negative regulation of transcription, DNA-templated
cardiac muscle cell differentiation
negative regulation of canonical Wnt signaling pathway
beta-catenin binding
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367725 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367723 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367726 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367718 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367724 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367722 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367719 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367720 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P470311 | AK003719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P470311 | NM_028634.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P470311 | AK003719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P470311 | NM_028634.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32304 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17381 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154715 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P470311 | AK003719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P470311 | NM_028634.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P470311 | AK003719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P470311 | NM_028634.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30367766 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154715 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08430899 | - | - |
| Whole Mouse Genome G4122A | A_51_P470311 | AK003719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P470311 | NM_028634.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P470311 | AK003719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P470311 | NM_028634.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127962 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12359611 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12359602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12359610 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12359608 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12359601 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12359604 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12359612 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367726 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367724 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367725 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367722 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367720 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367718 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367719 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30367723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30367698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12272630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11988629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12416076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12162173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12307681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12422953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12428830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12416073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12422950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11987035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12307688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12416070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12307696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30367700 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10425294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425298 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425296 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451305_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10425293 | - | - |
| Mouse Genome 430 2.0 Array | 1451305_at | - | - |
| Mouse Genome 430 2.0 Array | 1451305_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73739_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73739_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500005077.mm.1 | NM_028634.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005077.mm.1 | NM_028634.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005077.mm.1 | TC1500000677.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451305_at | - | - |
| Mouse Expression Set 430 | 1451305_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451305_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451305_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451305_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451305_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6200725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2640292 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640292 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2760937 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215039 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215039 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2760937 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002514.1_41-S | - | - |
| mouse-6 v1.1 (Illumina) | scl47741.5.1_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200725 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640292 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47741.5.1_17-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002514.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200725 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640292 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2760937 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2760937 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215039 | - | - |

Cby1
Genomic Map