Assay Details
| Gene Symbol: Dmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278226.1 | NP_001265155.1 | 4 - 5 | 424 | 98 | |
| NM_010059.3 | NP_034189.1 | 4 - 5 | 424 | 98 | ||
| NR_103477.1 | 4 - 5 | 424 | 98 | |||
| GenBank mRNA | AK076777.1 | - | 4 - 5 | 415 | 98 | |
| AK169570.1 | - | 4 - 5 | 424 | 98 | ||
| BC116767.1 | - | 4 - 5 | 309 | 40126086 | 98 | |
| BC119081.1 | - | 4 - 5 | 309 | 8733830 | 98 | |
| D58419.1 | - | 4 - 5 | 409 | 98 | ||
| D64107.1 | - | 4 - 5 | 398 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dmc1
Genomic Map