Assay Details
| Gene Symbol: Tmem184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK170086.1 | - | 1 - 2 | 100 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK170086.1 transcripts.
Set Membership: |
|

Tmem184b
Genomic Map