| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500014639.mm.1 |
NM_001253817.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014639.mm.1 |
NM_001253817.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014639.mm.1 |
NM_001253819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014639.mm.1 |
NM_001253819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014639.mm.1 |
NM_172608.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014639.mm.1 |
NM_172608.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014639.mm.1 |
TC1500001870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014638.mm.1 |
NM_001253817.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014638.mm.1 |
NM_001253817.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014638.mm.1 |
NM_001253819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014638.mm.1 |
NM_001253819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014638.mm.1 |
NM_172608.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014638.mm.1 |
NM_172608.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014638.mm.1 |
TC1500001870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014643.mm.1 |
NM_172608.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014643.mm.1 |
NM_172608.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014643.mm.1 |
TC1500001870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014640.mm.1 |
NM_001253817.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014640.mm.1 |
NM_001253817.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014640.mm.1 |
TC1500001870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014641.mm.1 |
NM_001253819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014641.mm.1 |
NM_001253819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014641.mm.1 |
NM_001253820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014641.mm.1 |
NM_001253820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014641.mm.1 |
NM_172608.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014641.mm.1 |
NM_172608.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014641.mm.1 |
TC1500001870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500014642.mm.1 |
NM_001253819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014642.mm.1 |
NM_001253819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014642.mm.1 |
NM_172608.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014642.mm.1 |
NM_172608.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014642.mm.1 |
TC1500001870.mm.1 |
MTA TC |