Assay Details
| Gene Symbol: Tmem184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253817.1 | NP_001240746.1 | 4 - 5 | 580 | 65 | |
| NM_001253819.1 | NP_001240748.1 | 4 - 5 | 601 | 65 | ||
| NM_001253820.1 | NP_001240749.1 | 2 - 3 | 146 | 65 | ||
| NM_172608.1 | NP_766196.1 | 4 - 5 | 626 | 65 | ||
| GenBank mRNA | AK028196.1 | - | 4 - 5 | 584 | 65 | |
| AK029129.1 | - | 4 - 5 | 604 | 65 | ||
| AK048138.1 | - | 4 - 5 | 580 | 65 | ||
| AK149366.1 | - | 4 - 5 | 607 | 65 | ||
| AK158789.1 | - | 4 - 5 | 627 | 65 | ||
| AK170086.1 | - | 4 - 5 | 604 | 65 | ||
| BC046959.1 | - | 4 - 5 | 580 | 6314456 | 65 | |
| CX239857.1 | - | 2 - 3 | 146 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001253820.1 transcripts.
Set Membership: |
|

Tmem184b
Genomic Map