Assay Details
| Gene Symbol: Tmem184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253817.1 | NP_001240746.1 | 7 - 8 | 921 | 78 | |
| NM_001253819.1 | NP_001240748.1 | 7 - 8 | 942 | 78 | ||
| NM_001253820.1 | NP_001240749.1 | 5 - 6 | 487 | 78 | ||
| NM_172608.1 | NP_766196.1 | 7 - 8 | 967 | 78 | ||
| GenBank mRNA | AK028196.1 | - | 7 - 8 | 925 | 78 | |
| AK029129.1 | - | 7 - 8 | 945 | 78 | ||
| AK048138.1 | - | 7 - 8 | 921 | 78 | ||
| AK149366.1 | - | 7 - 8 | 948 | 78 | ||
| AK158789.1 | - | 7 - 8 | 968 | 78 | ||
| AK170086.1 | - | 7 - 8 | 945 | 78 | ||
| AK187130.1 | - | 1 - 2 | 152 | 78 | ||
| BC046959.1 | - | 7 - 8 | 921 | 6314456 | 78 | |
| CX239857.1 | - | 5 - 6 | 487 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem184b
Genomic Map