Assay Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277255.1 | NP_001264184.1 | 6 - 7 | 810 | 108 | |
| NM_001277257.1 | NP_001264186.1 | 5 - 6 | 749 | 108 | ||
| NM_001277258.1 | NP_001264187.1 | 5 - 6 | 718 | 108 | ||
| NM_001277259.1 | NP_001264188.1 | 6 - 7 | 805 | 108 | ||
| NM_009612.3 | NP_033742.2 | 5 - 6 | 787 | 108 | ||
| GenBank mRNA | AK018754.1 | - | 2 - 3 | 289 | 108 | |
| AK134712.1 | - | 3 - 4 | 544 | 108 | ||
| AK160915.1 | - | 6 - 7 | 778 | 108 | ||
| AK170870.1 | - | 3 - 4 | 543 | 108 | ||
| BC014291.1 | - | 4 - 5 | 706 | 3982004 | 108 | |
| BC015083.1 | - | 4 - 5 | 668 | 3980151 | 108 | |
| CT010330.1 | - | 3 - 4 | 515 | 108 | ||
| L48015.1 | - | 5 - 6 | 789 | 108 | ||
| Z31664.1 | - | 5 - 6 | 755 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvrl1
Genomic Map