Assay Details
| Gene Symbol: Nr4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010444.2 | NP_034574.1 | 3 - 4 | 1145 | 57 | |
| GenBank mRNA | AK004709.1 | - | 3 - 4 | 1139 | 57 | |
| AK157203.1 | - | 3 - 4 | 1145 | 57 | ||
| BC004770.1 | - | 3 - 4 | 1125 | 3156434 | 57 | |
| CT010287.1 | - | 2 - 3 | 1017 | 57 | ||
| J04113.1 | - | 3 - 4 | 1128 | 57 | ||
| X60132.1 | - | 3 - 4 | 1095 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr4a1
Genomic Map