Assay Details
| Gene Symbol: Cela1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033612.2 | NP_291090.2 | 3 - 4 | 250 | 70 | |
| GenBank mRNA | AK007392.1 | - | 3 - 4 | 248 | 70 | |
| BC011218.1 | - | 3 - 4 | 224 | 4208442 | 70 | |
| CB951068.1 | - | 3 - 4 | 250 | 70 | ||
| M27347.1 | - | 2 - 3 | 133 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cela1
Genomic Map