Assay Details
| Gene Symbol: Cela1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033612.2 | NP_291090.2 | 6 - 7 | 660 | 80 | |
| GenBank mRNA | AK007392.1 | - | 6 - 7 | 658 | 80 | |
| BC011218.1 | - | 6 - 7 | 634 | 4208442 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cela1
Genomic Map