Assay Details
| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080981.1 | NP_001074450.1 | 12 - 13 | 1619 | 94 | |
| GenBank mRNA | AK014820.1 | - | 9 - 10 | 2680 | 94 | |
| BC049928.1 | - | 10 - 11 | 2349 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161535 | NM_001080981.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576937 | AK005297.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576937 | BC087890.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576937 | NM_025355.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17978 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110780 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102482 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161535 | NM_001080981.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102482 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110780 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08476787 | - | - |
| Whole Mouse Genome G4122A | A_52_P9347 | BC049928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9347 | AK014820.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9347 | NM_001080981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133327 | BC049928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133327 | AK014820.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110780 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102482 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12200027 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12200030 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11994556 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11994554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30475612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12126063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12368557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12384333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12073240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12139585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12200030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12368566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12120221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12427145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11994554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11910496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30475761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12200028 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432222 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432218 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432219 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432216 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432217 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439952_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430050_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432215 | - | - |
| Mouse Genome 430 2.0 Array | 1430050_at | - | - |
| Mouse Genome 430 2.0 Array | 1430050_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500017746.mm.1 | NM_001080981.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017746.mm.1 | NM_001080981.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500017746.mm.1 | TC1500002182.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430050_at | - | - |
| Mouse Expression Set 430 | 1430050_PM_at | - | - |
| Murine Genome U74v2 Set | 167109_PM_at | - | - |
| Murine Genome U74v2 Set | 167109_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430050_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439952_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430050_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439952_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 615844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103870014 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2525204 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2476999 | - | - |

Ddx23
Genomic Map