Assay Details
| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080981.1 | NP_001074450.1 | 15 - 16 | 2127 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx23| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080981.1 | NP_001074450.1 | 15 - 16 | 2127 | 63 | |
Set Membership: |
|