Assay Details
| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080981.1 | NP_001074450.1 | 1 - 2 | 68 | 69 | |
| GenBank mRNA | AK014820.1 | - | 1 - 2 | 61 | 69 | |
| BC049928.1 | - | 1 - 2 | 63 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001080981.1 transcripts.
Set Membership: |
|

Ddx23
Genomic Map