Assay Details
| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080981.1 | NP_001074450.1 | 4 - 5 | 484 | 90 | |
| GenBank mRNA | BC049928.1 | - | 4 - 5 | 479 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx23
Genomic Map