Assay Details
| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080981.1 | NP_001074450.1 | 8 - 9 | 936 | 75 | |
| GenBank mRNA | AK014820.1 | - | 6 - 7 | 1694 | 75 | |
| BC049928.1 | - | 6 - 7 | 1666 | 75 | ||
More Information
Set Membership: |
|

Ddx23
Genomic Map