Assay Details
| Gene Symbol: Tuba1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011653.2 | NP_035783.1 | 1 - 2 | 46 | 75 | |
| GenBank mRNA | AK077529.1 | - | 1 - 2 | 58 | 75 | |
| AK145404.1 | - | 1 - 2 | 95 | 75 | ||
| AK146983.1 | - | 1 - 2 | 95 | 75 | ||
| AK164335.1 | - | 1 - 2 | 97 | 75 | ||
| AK167687.1 | - | 1 - 2 | 95 | 75 | ||
| AK168762.1 | - | 1 - 2 | 95 | 75 | ||
| AK169610.1 | - | 1 - 2 | 96 | 75 | ||
| BC056169.1 | - | 1 - 2 | 57 | 5715646 | 75 | |
| BC083343.1 | - | 1 - 2 | 70 | 5687414 | 75 | |
| BC083344.1 | - | 1 - 2 | 65 | 5693701 | 75 | |
| BC083345.1 | - | 1 - 2 | 80 | 6415067 | 75 | |
| BC085256.1 | - | 1 - 2 | 79 | 6306481 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077529.1, AK145404.1, AK146983.1, AK164335.1, AK167687.1, AK168762.1, AK169610.1, BC056169.1, BC083343.1, BC083344.1, BC083345.1, BC085256.1, NM_011653.2 transcripts.
Set Membership: |
|

Tuba1a
Genomic Map