Assay Details
| Gene Symbol: Cox14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183256.3 | NP_899079.1 | 1 - 2 | 226 | 50 | |
| GenBank mRNA | AK002816.1 | - | 1 - 2 | 89 | 50 | |
| AK004378.1 | - | 1 - 2 | 45 | 50 | ||
| AK009401.1 | - | 1 - 2 | 198 | 50 | ||
| BC059905.1 | - | 1 - 2 | 178 | 6823331 | 50 | |
| BC092142.1 | - | 1 - 2 | 127 | 6814201 | 50 | |
| BP774190.1 | - | 1 - 2 | 253 | 50 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002816.1,AK004378.1,AK009401.1,BC059905.1,BC092142.1,NM_183256.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30481999 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30481997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30482002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30481998 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30482000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30482001 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | BP774190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | BU700473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | NM_183256.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | AK009401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | BC059905.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | BP774190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | BU700473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | NM_183256.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | AK009401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291839 | BC059905.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345809 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110889 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | BP774190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | BU700473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | NM_183256.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | AK009401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | BC059905.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | BP774190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | BU700473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | NM_183256.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | AK009401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291839 | BC059905.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30481997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30481995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30481993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30482000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30481999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30481996 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110889 | - | - |
| Whole Mouse Genome G4122A | A_51_P291839 | BP774190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | BU700473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | NM_183256.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | AK009401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | BC059905.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | BP774190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | BU700473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | NM_183256.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | AK009401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291839 | BC059905.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125224 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110889 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11891145 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11891155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11946144 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11891149 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30482001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30481997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30481998 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30481999 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30482000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30482002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30482003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30481946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30481996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30481997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11932855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30481993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11891144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30481995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30482014 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426824 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455006_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426822 | - | - |
| Mouse Genome 430 2.0 Array | 1455006_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455006_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66379_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66379_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500008091.mm.1 | NM_183256.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008091.mm.1 | NM_183256.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008091.mm.1 | TC1500001024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008092.mm.1 | NM_183256.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008092.mm.1 | NM_183256.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008092.mm.1 | TC1500001024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008095.mm.1 | NM_183256.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008095.mm.1 | NM_183256.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008095.mm.1 | TC1500001024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008090.mm.1 | NM_183256.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008090.mm.1 | NM_183256.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008090.mm.1 | TC1500001024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008093.mm.1 | NM_183256.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008093.mm.1 | NM_183256.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008093.mm.1 | TC1500001024.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455006_PM_at | - | - |
| Mouse Expression Set 430 | 1455006_at | - | - |
| Murine Genome U74v2 Set | 100537_at | - | - |
| Murine Genome U74v2 Set | 100537_PM_at | - | - |
| Murine Genome U74v2 Set | 162301_f_at | - | - |
| Murine Genome U74v2 Set | 162301_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455006_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455006_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455006_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455006_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 592095 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1400161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249408 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249408 | - | - |
| mouse-6 v1.1 (Illumina) | scl47524.3.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47524.3.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1249408 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47524.3.1_1-S | - | - |

Cox14
Genomic Map