Assay Details
| Gene Symbol: C230037L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077233.1 | 4 - 5 | 435 | 93 | ||
| GenBank mRNA | AK082325.1 | - | 3 - 4 | 1453 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | arylsulfatase A | Arsa |
AS-A, ASA, AW212749, As-2, As2, TISP73 |
-446.0 | Anti-sense | Chr. 15 - 89472476 - 89477424 on NCBI Build GRCm38 |
| Downstream | SH3/ankyrin domain gene 3 | Shank3 |
AI841104, SHANK3c-3, SHANK3c-4, Shank3b |
14.9kb | Sense | Chr. 15 - 89499707 - 89560261 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C230037L18Rik
Genomic Map