Assay Details
| Gene Symbol: Fkbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010220.4 | NP_034350.1 | 6 - 7 | 888 | 76 | |
| GenBank mRNA | AK169795.1 | - | 6 - 7 | 829 | 76 | |
| BC015260.1 | - | 6 - 7 | 827 | 4237766 | 76 | |
| BU613862.1 | - | 4 - 5 | 555 | 76 | ||
| CT010372.1 | - | 5 - 6 | 669 | 76 | ||
| U16959.1 | - | 6 - 7 | 888 | 76 | ||
| U36220.1 | - | 6 - 7 | 816 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp5
Genomic Map