Assay Details
| Gene Symbol: Fkbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010220.4 | NP_034350.1 | 9 - 10 | 1241 | 75 | |
| GenBank mRNA | AK169795.1 | - | 9 - 10 | 1182 | 75 | |
| BC015260.1 | - | 9 - 10 | 1180 | 4237766 | 75 | |
| CF949740.1 | - | 2 - 3 | 270 | 75 | ||
| CT010372.1 | - | 8 - 9 | 1022 | 75 | ||
| U16959.1 | - | 9 - 10 | 1241 | 75 | ||
| U36220.1 | - | 9 - 10 | 1169 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp5
Genomic Map