Assay Details
| Gene Symbol: Tcp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085555.1 | NP_001079024.1 | 4 - 5 | 409 | 91 | |
| NM_013687.3 | NP_038715.2 | 4 - 5 | 646 | 91 | ||
| GenBank mRNA | BC016610.1 | - | 4 - 5 | 629 | 91 | |
| BC058793.1 | - | 3 - 4 | 585 | 91 | ||
| BC089527.1 | - | 4 - 5 | 647 | 91 | ||
| BC089528.1 | - | 4 - 5 | 392 | 6741317 | 91 | |
| X52127.1 | - | 2 - 3 | 168 | 91 | ||
| X52128.1 | - | 4 - 5 | 624 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcp11
Genomic Map