Assay Details
| Gene Symbol: Tcp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085555.1 | NP_001079024.1 | 7 - 8 | 980 | 97 | |
| NM_013687.3 | NP_038715.2 | 7 - 8 | 1217 | 97 | ||
| GenBank mRNA | BC016610.1 | - | 7 - 8 | 1200 | 97 | |
| BC058793.1 | - | 6 - 7 | 1156 | 97 | ||
| BC089527.1 | - | 7 - 8 | 1218 | 97 | ||
| BC089528.1 | - | 7 - 8 | 963 | 6741317 | 97 | |
| X52127.1 | - | 5 - 6 | 739 | 97 | ||
| X52128.1 | - | 7 - 8 | 1195 | 97 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tcp11
Genomic Map