Assay Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029624.4 | NP_083900.3 | 3 - 4 | 527 | 78 | |
| GenBank mRNA | AK010312.1 | - | 3 - 4 | 498 | 78 | |
| AK053241.1 | - | 3 - 4 | 504 | 78 | ||
| AK154622.1 | - | 3 - 4 | 524 | 78 | ||
| AK169983.1 | - | 3 - 4 | 525 | 78 | ||
| AK172374.1 | - | 3 - 4 | 506 | 78 | ||
| BC020104.1 | - | 3 - 4 | 534 | 3963699 | 78 | |
More Information
Set Membership: |
|

Lmf1
Genomic Map