Assay Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029624.4 | NP_083900.3 | 4 - 5 | 674 | 78 | |
| NR_045520.1 | 2 - 3 | 353 | 78 | |||
| GenBank mRNA | AK010312.1 | - | 4 - 5 | 645 | 78 | |
| AK041659.1 | - | 2 - 3 | 341 | 78 | ||
| AK053241.1 | - | 4 - 5 | 651 | 78 | ||
| AK142235.1 | - | 4 - 5 | 706 | 78 | ||
| AK154622.1 | - | 4 - 5 | 671 | 78 | ||
| AK169983.1 | - | 4 - 5 | 672 | 78 | ||
| AK172374.1 | - | 4 - 5 | 653 | 78 | ||
| BC020104.1 | - | 4 - 5 | 681 | 3963699 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041659.1 transcripts.
Set Membership: |
|

Lmf1
Genomic Map