Assay Details
| Gene Symbol: Cacna1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163691.1 | NP_001157163.1 | 31 - 32 | 5794 | 88 | |
| NM_021415.4 | NP_067390.4 | 31 - 32 | 5794 | 88 | ||
| GenBank mRNA | AK122452.1 | - | 20 - 21 | 2767 | 88 | |
| AK135951.1 | - | 13 - 14 | 1541 | 88 | ||
| AK147242.1 | - | 31 - 32 | 5795 | 88 | ||
| BC138026.1 | - | 31 - 32 | 5633 | 8861045 | 88 | |
| BU612210.1 | - | 5 - 6 | 505 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cacna1h
Genomic Map