| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700015246.mm.1 |
NM_018857.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015246.mm.1 |
NM_018857.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015246.mm.1 |
TC1700001671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015244.mm.1 |
NM_018857.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015244.mm.1 |
NM_018857.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015244.mm.1 |
TC1700001671.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015245.mm.1 |
NM_018857.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015245.mm.1 |
NM_018857.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015245.mm.1 |
TC1700001671.mm.1 |
MTA TC |