Assay Details
| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163661.1 | NP_001157133.1 | 13 - 14 | 2227 | 85 | |
| NM_027880.2 | NP_082156.2 | 13 - 14 | 2138 | 85 | ||
| GenBank mRNA | AK004582.1 | - | 13 - 14 | 2008 | 85 | |
| AK028658.1 | - | 13 - 14 | 1992 | 85 | ||
| AK045321.1 | - | 6 - 7 | 1674 | 85 | ||
| AK050773.1 | - | 13 - 14 | 1957 | 85 | ||
| AK084830.1 | - | 13 - 14 | 2011 | 87 | ||
| AK129189.1 | - | 12 - 13 | 2300 | 85 | ||
| AK143827.1 | - | 13 - 14 | 2228 | 85 | ||
| AK146963.1 | - | 13 - 14 | 2014 | 85 | ||
| AK154727.1 | - | 13 - 14 | 2040 | 85 | ||
| AK158181.1 | - | 8 - 9 | 85 | |||
| BC011077.1 | - | 13 - 14 | 1988 | 3984911 | 85 | |
| BC080713.1 | - | 12 - 13 | 2247 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Telo2
Genomic Map