Assay Details
| Gene Symbol: Rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028009.3 | NP_082285.1 | 2 - 3 | 282 | 68 | |
| GenBank mRNA | AK049334.1 | - | 2 - 3 | 283 | 68 | |
| BC018186.1 | - | 2 - 3 | 243 | 4160116 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpusd1
Genomic Map