Assay Details
| Gene Symbol: Rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028009.3 | NP_082285.1 | 3 - 4 | 412 | 61 | |
| GenBank mRNA | AK009926.1 | - | 2 - 3 | 244 | 61 | |
| AK049334.1 | - | 3 - 4 | 413 | 61 | ||
| BC018186.1 | - | 3 - 4 | 373 | 4160116 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpusd1
Genomic Map