Assay Details
| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145409.2 | NP_663384.2 | 12 - 13 | 1635 | 123 | |
| GenBank mRNA | BC024142.1 | - | 12 - 13 | 1622 | 5149957 | 123 |
More Information
Updated Mapping Information
Set Membership: |
|

Chtf18
Genomic Map