Assay Details
| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145409.2 | NP_663384.2 | 18 - 19 | 2514 | 57 | |
| GenBank mRNA | AK052673.1 | - | 17 - 18 | 2725 | 57 | |
| AK077947.1 | - | 18 - 19 | 2481 | 57 | ||
| BC024142.1 | - | 18 - 19 | 2501 | 5149957 | 57 | |
| BC083096.1 | - | 15 - 16 | 3232 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chtf18
Genomic Map