Assay Details
| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145409.2 | NP_663384.2 | 20 - 21 | 2793 | 71 | |
| GenBank mRNA | AK052673.1 | - | 19 - 20 | 3004 | 71 | |
| AK077947.1 | - | 20 - 21 | 2760 | 71 | ||
| BC083096.1 | - | 17 - 18 | 3511 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chtf18
Genomic Map