Assay Details
| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145409.2 | NP_663384.2 | 3 - 4 | 498 | 64 | |
| GenBank mRNA | AK052673.1 | - | 3 - 4 | 457 | 64 | |
| AK077947.1 | - | 3 - 4 | 465 | 64 | ||
| AK181238.1 | - | 1 - 2 | 61 | 64 | ||
| BC024142.1 | - | 3 - 4 | 485 | 5149957 | 64 | |
| BC083096.1 | - | 3 - 4 | 461 | 64 | ||
More Information
Set Membership: |
|

Chtf18
Genomic Map