Assay Details
| Gene Symbol: Rhot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145999.2 | NP_666111.1 | 12 - 13 | 1366 | 56 | |
| GenBank mRNA | AK142183.1 | - | 9 - 10 | 1273 | 56 | |
| AY311390.1 | - | 12 - 13 | 1366 | 56 | ||
| BC023163.1 | - | 2 - 3 | 136 | 56 | ||
| BC029777.1 | - | 12 - 13 | 999 | 4950061 | 56 | |
| BC034062.1 | - | 12 - 13 | 1021 | 4207654 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhot2
Genomic Map