Assay Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011144.6 | NP_035274.2 | 1 - 2 | 319 | 92 | |
| GenBank mRNA | AK035676.1 | - | 1 - 2 | 320 | 92 | |
| BC016892.1 | - | 1 - 2 | 312 | 4222384 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035676.1, BC016892.1, NM_011144.6 transcripts.
Set Membership: |
|

Ppara
Genomic Map