Assay Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113418.1 | NP_001106889.1 | 6 - 7 | 919 | 70 | |
| NM_011144.6 | NP_035274.2 | 6 - 7 | 1157 | 70 | ||
| GenBank mRNA | AK035676.1 | - | 6 - 7 | 1158 | 70 | |
| AK081709.1 | - | 6 - 7 | 920 | 70 | ||
| AK149460.1 | - | 6 - 7 | 856 | 70 | ||
| BC016892.1 | - | 6 - 7 | 1150 | 4222384 | 70 | |
| X57638.1 | - | 6 - 7 | 878 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppara
Genomic Map