Assay Details
| Gene Symbol: Erbb2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005868.2 | NP_001005868.1 | 9 - 10 | 1066 | 76 | |
| NM_001289473.1 | NP_001276402.1 | 9 - 10 | 1066 | 76 | ||
| NM_001289474.1 | NP_001276403.1 | 9 - 10 | 1066 | 76 | ||
| NM_001289475.1 | NP_001276404.1 | 9 - 10 | 1066 | 76 | ||
| GenBank mRNA | AK122473.1 | - | 9 - 10 | 745 | 76 | |
| AK169529.1 | - | 9 - 10 | 995 | 76 | ||
| BC141178.1 | - | 9 - 10 | 775 | 9055790 | 76 | |
| BC141188.1 | - | 9 - 10 | 775 | 9055800 | 76 | |
| BC145498.1 | - | 9 - 10 | 775 | 9054061 | 76 | |
| BC145509.1 | - | 9 - 10 | 775 | 9054072 | 76 | |
| EU233994.1 | - | 11 - 12 | 1822 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2ip
Genomic Map