Assay Details
| Gene Symbol: Erbb2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005868.2 | NP_001005868.1 | 15 - 16 | 1695 | 116 | |
| NM_001289473.1 | NP_001276402.1 | 15 - 16 | 1695 | 116 | ||
| NM_001289474.1 | NP_001276403.1 | 15 - 16 | 1695 | 116 | ||
| NM_001289475.1 | NP_001276404.1 | 15 - 16 | 1695 | 116 | ||
| GenBank mRNA | AK029054.1 | - | 4 - 5 | 377 | 116 | |
| AK122473.1 | - | 15 - 16 | 1374 | 116 | ||
| AK169529.1 | - | 15 - 16 | 1624 | 116 | ||
| BC141178.1 | - | 15 - 16 | 1404 | 9055790 | 116 | |
| BC141188.1 | - | 15 - 16 | 1404 | 9055800 | 116 | |
| BC145498.1 | - | 15 - 16 | 1404 | 9054061 | 116 | |
| BC145509.1 | - | 15 - 16 | 1404 | 9054072 | 116 | |
| EU233994.1 | - | 17 - 18 | 2452 | 116 | ||
More Information
Set Membership: |
|

Erbb2ip
Genomic Map