Assay Details
| Gene Symbol: Erbb2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005868.2 | NP_001005868.1 | 24 - 25 | 4473 | 92 | |
| NM_001289473.1 | NP_001276402.1 | 23 - 24 | 4329 | 92 | ||
| NM_001289475.1 | NP_001276404.1 | 23 - 24 | 4356 | 92 | ||
| GenBank mRNA | AF263743.1 | - | 3 - 4 | 1210 | 92 | |
| AK029054.1 | - | 13 - 14 | 3155 | 92 | ||
| AK051733.1 | - | 3 - 4 | 988 | 92 | ||
| AK122473.1 | - | 23 - 24 | 3930 | 92 | ||
| BC005691.1 | - | 3 - 4 | 454 | 92 | ||
| BC028256.1 | - | 4 - 5 | 1916 | 92 | ||
| BC141188.1 | - | 24 - 25 | 4182 | 9055800 | 92 | |
| BC145498.1 | - | 23 - 24 | 4065 | 9054061 | 92 | |
| BC145509.1 | - | 23 - 24 | 4038 | 9054072 | 92 | |
| EU233994.1 | - | 26 - 27 | 5423 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2ip
Genomic Map