Assay Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021790.2 | NP_068562.1 | 6 - 7 | 684 | 175 | |
| NM_181061.5 | NP_851406.1 | 6 - 7 | 807 | 175 | ||
| GenBank mRNA | AB043687.1 | - | 6 - 7 | 684 | 175 | |
| AK082939.1 | - | 6 - 7 | 464 | 175 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpk
Genomic Map