Assay Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021790.2 | NP_068562.1 | 8 - 9 | 900 | 62 | |
| NM_181061.5 | NP_851406.1 | 8 - 9 | 1023 | 62 | ||
| NR_075088.1 | 8 - 9 | 1036 | 62 | |||
| GenBank mRNA | AB043687.1 | - | 8 - 9 | 900 | 62 | |
| AK082939.1 | - | 8 - 9 | 680 | 62 | ||
| AK135773.1 | - | 416 | 62 | |||
| BC028655.1 | - | 8 - 9 | 1036 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cenpk
Genomic Map