Assay Details
| Gene Symbol: Ecel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277925.1 | NP_001264854.1 | 13 - 14 | 2078 | 69 | |
| NM_021306.3 | NP_067281.2 | 13 - 14 | 2070 | 69 | ||
| GenBank mRNA | AK039467.1 | - | 13 - 14 | 2071 | 69 | |
| AK159365.1 | - | 13 - 14 | 2080 | 69 | ||
| BC057569.1 | - | 12 - 13 | 1900 | 6413731 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecel1
Genomic Map