| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100022279.mm.1 |
NM_001277925.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022279.mm.1 |
TC0100002758.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022277.mm.1 |
NM_001277925.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022277.mm.1 |
NM_021306.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022277.mm.1 |
TC0100002758.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022275.mm.1 |
NM_001277925.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022275.mm.1 |
NM_021306.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022275.mm.1 |
TC0100002758.mm.1 |
MTA TC |