Assay Details
| Gene Symbol: Ecel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277925.1 | NP_001264854.1 | 3 - 4 | 1076 | 68 | |
| NM_021306.3 | NP_067281.2 | 3 - 4 | 1068 | 68 | ||
| GenBank mRNA | AB026294.1 | - | 2 - 3 | 896 | 68 | |
| AK039467.1 | - | 3 - 4 | 1068 | 68 | ||
| AK159365.1 | - | 3 - 4 | 1078 | 68 | ||
| BC057569.1 | - | 2 - 3 | 898 | 6413731 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecel1
Genomic Map